Search
Search

phf20b| Gene Symbol: phf20b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004114.1 | NP_001004114.1 | 6 - 7 | 1012 | 76 | |
| XM_005161993.4 | XP_005162050.1 | 6 - 7 | 1075 | 76 | ||
| GenBank mRNA | BC080250.1 | - | 6 - 7 | 1012 | 76 | |
Set Membership: |
|