Search
Search

dntt| Gene Symbol: dntt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014817.1 | NP_001014817.1 | 6 - 7 | 840 | 61 | |
| GenBank mRNA | AY518898.1 | - | 6 - 7 | 840 | 61 | |
| BC163775.1 | - | 6 - 7 | 870 | 61 | ||
Set Membership: |
|