Search
Search

fbxl14b| Gene Symbol: fbxl14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015043.2 | NP_001015043.1 | 1 | 546 | 62 | |
| GenBank mRNA | AF467462.1 | - | 1 | 315 | 62 | |
| BC163001.1 | - | 1 | 546 | 62 | ||
| BC163002.1 | - | 1 | 546 | 62 | ||
Set Membership: |
|