Search
Search

ankha| Gene Symbol: ankha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030259.2 | NP_001025430.2 | 11 - 12 | 1676 | 61 | |
| XM_021470216.1 | XP_021325891.1 | 9 - 10 | 1354 | 61 | ||
| GenBank mRNA | BC096801.1 | - | 11 - 12 | 1983 | 61 | |
Set Membership: |
|