Search
Search

pmelb| Gene Symbol: pmelb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033918.2 | NP_001029090.2 | 10 - 11 | 1652 | 62 | |
| GenBank mRNA | AY864065.1 | - | 10 - 11 | 1622 | 62 | |
| BC116462.1 | - | 10 - 11 | 1652 | 62 | ||
Set Membership: |
|