Search
Search

pmelb| Gene Symbol: pmelb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033918.2 | NP_001029090.2 | 2 - 3 | 200 | 67 | |
| XM_005162378.4 | XP_005162435.1 | 2 - 3 | 242 | 67 | ||
| GenBank mRNA | AY864065.1 | - | 2 - 3 | 170 | 67 | |
| BC116462.1 | - | 2 - 3 | 200 | 67 | ||
| KM655726.1 | - | 2 - 3 | 170 | 67 | ||
Set Membership: |
|