Search
Search

scn4aa| Gene Symbol: scn4aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039825.1 | NP_001034914.1 | 12 - 13 | 1858 | 133 | |
| XM_017358479.2 | XP_017213968.1 | 13 - 14 | 2044 | 133 | ||
| GenBank mRNA | DQ149506.1 | - | 12 - 13 | 1858 | 133 | |
Set Membership: |
|