Search
Search

ssuh2rs1| Gene Symbol: ssuh2rs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076658.1 | NP_001070126.1 | 4 - 5 | 396 | 118 | |
| XM_005166701.4 | XP_005166758.1 | 5 - 6 | 570 | 118 | ||
| GenBank mRNA | BC124353.1 | - | 4 - 5 | 396 | 118 | |
Set Membership: |
|