Search
Search

senp3b| Gene Symbol: senp3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083842.1 | NP_001077311.1 | 10 - 11 | 1723 | 75 | |
| GenBank mRNA | BC096903.1 | - | 9 - 10 | 878 | 75 | |
| BC124470.1 | - | 9 - 10 | 1293 | 75 | ||
| BC134841.1 | - | 10 - 11 | 1723 | 75 | ||
| BC153650.1 | - | 9 - 10 | 941 | 75 | ||
Set Membership: |
|