Search
Search

pax2a| Gene Symbol: pax2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131184.2 | NP_571259.1 | 7 - 8 | 1411 | 80 | |
| XM_005156744.4 | XP_005156801.1 | 8 - 9 | 2099 | 80 | ||
| XM_005156747.4 | XP_005156804.1 | 8 - 9 | 2092 | 80 | ||
| XM_005156748.4 | XP_005156805.1 | 7 - 8 | 2048 | 80 | ||
| XM_021480502.1 | XP_021336177.1 | 9 - 10 | 2760 | 80 | ||
| XM_021480503.1 | XP_021336178.1 | 9 - 10 | 2758 | 80 | ||
| XM_021480504.1 | XP_021336179.1 | 9 - 10 | 2219 | 80 | ||
| XM_021480505.1 | XP_021336180.1 | 8 - 9 | 2112 | 80 | ||
| XM_021480506.1 | XP_021336181.1 | 8 - 9 | 2097 | 80 | ||
| XM_021480507.1 | XP_021336182.1 | 8 - 9 | 2105 | 80 | ||
| XM_021480508.1 | XP_021336183.1 | 7 - 8 | 2061 | 80 | ||
| XM_021480509.1 | XP_021336184.1 | 7 - 8 | 2054 | 80 | ||
| GenBank mRNA | AF067530.1 | - | 8 - 9 | 952 | 80 | |
| AF067531.1 | - | 7 - 8 | 907 | 80 | ||
| AF067534.1 | - | 8 - 9 | 964 | 80 | ||
| AF067537.1 | - | 8 - 9 | 964 | 80 | ||
| AF067538.1 | - | 7 - 8 | 919 | 80 | ||
| AF067539.1 | - | 7 - 8 | 907 | 80 | ||
| AF067540.1 | - | 8 - 9 | 952 | 80 | ||
| AF067541.1 | - | 7 - 8 | 919 | 80 | ||
| BC066690.1 | - | 7 - 8 | 1411 | 80 | ||
| X63961.1 | - | 1533 | 80 | |||
Set Membership: |
|