Search
Search

spon1b| Gene Symbol: spon1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131517.1 | NP_571592.1 | 12 - 13 | 1831 | 98 | |
| XM_005174346.4 | XP_005174403.1 | 12 - 13 | 2127 | 98 | ||
| GenBank mRNA | AB006087.1 | - | 12 - 13 | 1831 | 98 | |
| BC162653.1 | - | 12 - 13 | 1685 | 98 | ||
| KM655691.1 | - | 12 - 13 | 1652 | 98 | ||
Set Membership: |
|