Search
Search

coro2a| Gene Symbol: coro2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199643.2 | NP_955937.1 | 9 - 10 | 1181 | 67 | |
| XM_005159876.4 | XP_005159933.1 | 10 - 11 | 1416 | 67 | ||
| XM_005159877.4 | XP_005159934.1 | 10 - 11 | 1437 | 67 | ||
| GenBank mRNA | BC060941.1 | - | 9 - 10 | 1185 | 67 | |
Set Membership: |
|