Search
Search

nuf2| Gene Symbol: nuf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200310.1 | NP_956604.1 | 9 - 10 | 904 | 59 | |
| XM_005155671.3 | XP_005155728.1 | 10 - 11 | 890 | 59 | ||
| GenBank mRNA | BC050181.1 | - | 9 - 10 | 904 | 59 | |
Set Membership: |
|