Search
Search

cdc14aa| Gene Symbol: cdc14aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201149.1 | NP_957443.1 | 4 - 5 | 618 | 104 | |
| XM_005163259.4 | XP_005163316.1 | 4 - 5 | 680 | 104 | ||
| GenBank mRNA | BC055188.1 | - | 4 - 5 | 618 | 104 | |
Set Membership: |
|