Search
Search

ncln| Gene Symbol: ncln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207057.1 | NP_996940.1 | 5 - 6 | 786 | 62 | |
| XM_005161589.4 | XP_005161646.1 | 5 - 6 | 789 | 62 | ||
| XM_005161590.4 | XP_005161647.1 | 5 - 6 | 789 | 62 | ||
| XM_005161591.4 | XP_005161648.1 | 5 - 6 | 789 | 62 | ||
| GenBank mRNA | BC056523.1 | - | 5 - 6 | 779 | 62 | |
| BC066393.1 | - | 5 - 6 | 786 | 62 | ||
Set Membership: |
|