Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ilf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212599.1 | NP_997764.1 | 7 - 8 | 850 | 68 | |
XM_005163739.4 | XP_005163796.1 | 7 - 8 | 922 | 68 | ||
XM_005163740.4 | XP_005163797.1 | 7 - 8 | 922 | 68 | ||
XM_005163741.3 | XP_005163798.1 | 7 - 8 | 919 | 68 | ||
XM_005163742.4 | XP_005163799.1 | 7 - 8 | 919 | 68 | ||
XM_009299225.3 | XP_009297500.1 | 6 - 7 | 830 | 68 | ||
GenBank mRNA | BC067172.1 | - | 7 - 8 | 850 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |