Search
Search

acp5a| Gene Symbol: acp5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214773.1 | NP_999938.1 | 5 - 6 | 811 | 77 | |
| XM_005165725.4 | XP_005165782.1 | 5 - 6 | 819 | 77 | ||
| XM_005165726.4 | XP_005165783.1 | 5 - 6 | 896 | 77 | ||
| GenBank mRNA | BC055256.1 | - | 5 - 6 | 811 | 77 | |
Set Membership: |
|