Search
Search

fn1b| Gene Symbol: fn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013261.1 | NP_001013279.1 | 36 - 37 | 5832 | 112 | |
| XM_005160284.3 | XP_005160341.1 | 36 - 37 | 5864 | 112 | ||
| XM_009305554.3 | XP_009303829.1 | 37 - 38 | 6144 | 112 | ||
| GenBank mRNA | AY538257.1 | - | 36 - 37 | 5832 | 112 | |
| AY725818.1 | - | 37 - 38 | 6123 | 112 | ||
Set Membership: |
|