Search
Search

btbd7| Gene Symbol: btbd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025551.1 | NP_001020722.1 | 9 - 10 | 2577 | 72 | |
| XM_005160586.3 | XP_005160643.1 | 10 - 11 | 3146 | 72 | ||
Set Membership: |
|