Search
Search

tnikb| Gene Symbol: tnikb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037676.2 | NP_001032765.2 | 24 - 25 | 3083 | 90 | |
| XM_005162778.4 | XP_005162835.1 | 24 - 25 | 3443 | 90 | ||
| XM_005162779.4 | XP_005162836.1 | 23 - 24 | 3088 | 90 | ||
| XM_005162780.4 | XP_005162837.1 | 22 - 23 | 3001 | 90 | ||
| XM_021470183.1 | XP_021325858.1 | 26 - 27 | 3476 | 90 | ||
| XM_021470184.1 | XP_021325859.1 | 25 - 26 | 3389 | 90 | ||
| XM_021470186.1 | XP_021325861.1 | 26 - 27 | 3477 | 90 | ||
| XM_021470187.1 | XP_021325862.1 | 25 - 26 | 3121 | 90 | ||
| XM_021470188.1 | XP_021325863.1 | 24 - 25 | 3034 | 90 | ||
| GenBank mRNA | BC108020.1 | - | 5 - 6 | 698 | 90 | |
Set Membership: |
|