Search
Search

spint2| Gene Symbol: spint2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077250.2 | NP_001070718.2 | 1 - 2 | 485 | 64 | |
| GenBank mRNA | BC115335.1 | - | 1 - 2 | 242 | 64 | |
| BC124739.1 | - | 1 - 2 | 445 | 64 | ||
| EB963426.1 | - | 1 - 2 | 489 | 64 | ||
| EH457273.1 | - | 1 - 2 | 492 | 64 | ||
Set Membership: |
|