Search
Search

msx3| Gene Symbol: msx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131272.2 | NP_571347.2 | 1 - 2 | 520 | 83 | |
| GenBank mRNA | BC071302.1 | - | 1 - 2 | 503 | 83 | |
| EH593082.1 | - | 1 - 2 | 525 | 83 | ||
| X65061.1 | - | 517 | 83 | |||
Set Membership: |
|