Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ldb2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131315.3 | NP_571390.1 | 1 - 2 | 269 | 70 | |
XM_009307714.3 | XP_009305989.1 | 1 - 2 | 296 | 70 | ||
XM_009307716.3 | XP_009305991.1 | 1 - 2 | 296 | 70 | ||
GenBank mRNA | AF031377.1 | - | 1 - 2 | 254 | 70 | |
BC090063.1 | - | 1 - 2 | 269 | 70 | ||
BC164481.1 | - | 1 - 2 | 164 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |