Search
Search

rps18| Gene Symbol: rps18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173234.1 | NP_775341.1 | 3 - 4 | 320 | 144 | |
| GenBank mRNA | AF210641.1 | - | 3 - 4 | 225 | 144 | |
| AY099517.1 | - | 3 - 4 | 320 | 144 | ||
| BC062289.1 | - | 3 - 4 | 301 | 144 | ||
Set Membership: |
|