Search
Search

unc50| Gene Symbol: unc50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200247.1 | NP_956541.1 | 4 - 5 | 670 | 93 | |
| GenBank mRNA | BC047831.1 | - | 4 - 5 | 670 | 93 | |
| BC164589.1 | - | 3 - 4 | 561 | 93 | ||
Set Membership: |
|