Search
Search

abhd2a| Gene Symbol: abhd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200914.1 | NP_957208.1 | 9 - 10 | 1266 | 70 | |
| GenBank mRNA | BC047183.1 | - | 9 - 10 | 1266 | 70 | |
| BC165808.1 | - | 7 - 8 | 1034 | 70 | ||
Set Membership: |
|