Search
Search

rhot1a| Gene Symbol: rhot1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212704.1 | NP_997869.1 | 16 - 17 | 1720 | 134 | |
| XM_009299425.3 | XP_009297700.1 | 16 - 17 | 1749 | 134 | ||
| XM_009299426.3 | XP_009297701.1 | 16 - 17 | 1748 | 134 | ||
| XM_009299427.3 | XP_009297702.1 | 16 - 17 | 1749 | 134 | ||
| GenBank mRNA | BC044431.1 | - | 16 - 17 | 1715 | 134 | |
| BC068190.1 | - | 16 - 17 | 1720 | 134 | ||
Set Membership: |
|