Search
Search

prpf18| Gene Symbol: prpf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002204.1 | NP_001002204.1 | 3 - 4 | 344 | 68 | |
| XM_021467629.1 | XP_021323304.1 | 4 - 5 | 460 | 68 | ||
| GenBank mRNA | BC074081.1 | - | 3 - 4 | 344 | 68 | |
Set Membership: |
|