Search
Search

asb5b| Gene Symbol: asb5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017753.1 | NP_001017753.1 | 6 - 7 | 898 | 84 | |
| XM_005159984.4 | XP_005160041.1 | 6 - 7 | 903 | 84 | ||
| XM_009292651.2 | XP_009290926.1 | 7 - 8 | 814 | 84 | ||
| XM_021474745.1 | XP_021330420.1 | 5 - 6 | 803 | 84 | ||
| XM_021474750.1 | XP_021330425.1 | 5 - 6 | 797 | 84 | ||
| GenBank mRNA | BC093197.1 | - | 6 - 7 | 898 | 84 | |
Set Membership: |
|