Search
Search

flvcr1| Gene Symbol: flvcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025522.1 | NP_001020693.1 | 6 - 7 | 1206 | 70 | |
| XM_005160739.3 | XP_005160796.1 | 6 - 7 | 1633 | 70 | ||
| XM_009294763.3 | XP_009293038.1 | 7 - 8 | 1514 | 70 | ||
Set Membership: |
|