Search
Search

lgi3| Gene Symbol: lgi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039679.1 | NP_001034768.1 | 3 - 4 | 457 | 65 | |
| GenBank mRNA | AY309263.1 | - | 3 - 4 | 457 | 65 | |
| BC162727.1 | - | 3 - 4 | 488 | 65 | ||
Set Membership: |
|