Search
Search

kcnh2a| Gene Symbol: kcnh2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042722.2 | NP_001036187.2 | 14 - 15 | 3820 | 115 | |
| XM_021480879.1 | XP_021336554.1 | 14 - 15 | 3798 | 115 | ||
| XM_021480881.1 | XP_021336556.1 | 10 - 11 | 3227 | 115 | ||
| GenBank mRNA | DQ022613.1 | - | 14 - 15 | 3360 | 115 | |
Set Membership: |
|