Search
Search

wnt2bb| Gene Symbol: wnt2bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044344.1 | NP_001037809.1 | 3 - 4 | 915 | 92 | |
| GenBank mRNA | BC162931.1 | - | 3 - 4 | 941 | 92 | |
| BC162939.1 | - | 3 - 4 | 941 | 92 | ||
| DQ231559.1 | - | 3 - 4 | 915 | 92 | ||
Set Membership: |
|