Search
Search

crtac1a| Gene Symbol: crtac1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080178.1 | NP_001073647.1 | 8 - 9 | 1145 | 82 | |
| XM_005160175.4 | XP_005160232.1 | 8 - 9 | 1227 | 82 | ||
| XM_021475056.1 | XP_021330731.1 | 8 - 9 | 1228 | 82 | ||
| XM_021475062.1 | XP_021330737.1 | 8 - 9 | 1196 | 82 | ||
| XM_021475068.1 | XP_021330743.1 | 8 - 9 | 1226 | 82 | ||
| GenBank mRNA | BC146702.1 | - | 8 - 9 | 1166 | 82 | |
| DQ351708.1 | - | 8 - 9 | 1145 | 82 | ||
Set Membership: |
|