Search
Search

ncaph| Gene Symbol: ncaph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080196.1 | NP_001073665.1 | 16 - 17 | 2045 | 76 | |
| XM_021476302.1 | XP_021331977.1 | 16 - 17 | 2073 | 76 | ||
| GenBank mRNA | BC129325.1 | - | 16 - 17 | 2045 | 76 | |
| BC165645.1 | - | 15 - 16 | 1970 | 76 | ||
Set Membership: |
|