Search
Search

smn1| Gene Symbol: smn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131191.1 | NP_571266.1 | 4 - 5 | 509 | 68 | |
| GenBank mRNA | BC060942.1 | - | 4 - 5 | 490 | 68 | |
| Y17256.1 | - | 509 | 68 | |||
Set Membership: |
|