Search
Search

sfrp5| Gene Symbol: sfrp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131858.1 | NP_571933.1 | 2 - 3 | 807 | 89 | |
| GenBank mRNA | AY050560.1 | - | 2 - 3 | 684 | 89 | |
| BC085455.1 | - | 2 - 3 | 807 | 89 | ||
| BC164265.1 | - | 2 - 3 | 641 | 89 | ||
Set Membership: |
|