Search
Search

thbs3a| Gene Symbol: thbs3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173225.1 | NP_775332.1 | 1 - 2 | 178 | 61 | |
| GenBank mRNA | AF462310.1 | - | 1 - 2 | 178 | 61 | |
| BC085524.1 | - | 1 - 2 | 176 | 61 | ||
| BC165677.1 | - | 1 - 2 | 128 | 61 | ||
Set Membership: |
|