Search
Search

thbs3a| Gene Symbol: thbs3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173225.1 | NP_775332.1 | 3 - 4 | 636 | 96 | |
| GenBank mRNA | AF462310.1 | - | 3 - 4 | 636 | 96 | |
| BC085524.1 | - | 3 - 4 | 634 | 96 | ||
| BC165677.1 | - | 3 - 4 | 586 | 96 | ||
Set Membership: |
|