Search
Search

lgi1a| Gene Symbol: lgi1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199627.2 | NP_955921.2 | 5 - 6 | 629 | 83 | |
| GenBank mRNA | AY309261.1 | - | 5 - 6 | 571 | 83 | |
| BC056753.1 | - | 5 - 6 | 612 | 83 | ||
| BC065665.1 | - | 5 - 6 | 593 | 83 | ||
| CU468780.1 | - | 5 - 6 | 461 | 83 | ||
| CU638810.1 | - | 5 - 6 | 474 | 83 | ||
Set Membership: |
|