Search
Search

rpl10a| Gene Symbol: rpl10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199636.1 | NP_955930.1 | 3 - 4 | 181 | 81 | |
| XM_021478186.1 | XP_021333861.1 | 3 - 4 | 382 | 81 | ||
| GenBank mRNA | BC059454.1 | - | 3 - 4 | 181 | 81 | |
Set Membership: |
|