Search
Search

rpl34| Gene Symbol: rpl34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201122.1 | NP_957416.1 | 3 - 4 | 198 | 86 | |
| XM_005166535.3 | XP_005166592.1 | 3 - 4 | 419 | 86 | ||
| GenBank mRNA | BC049023.1 | - | 3 - 4 | 198 | 86 | |
| BC062280.1 | - | 2 - 3 | 182 | 86 | ||
Set Membership: |
|