Search
Search

lhfpl2a| Gene Symbol: lhfpl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205663.1 | NP_991226.1 | 2 - 3 | 625 | 85 | |
| XM_021468792.1 | XP_021324467.1 | 2 - 3 | 773 | 85 | ||
| GenBank mRNA | BC065671.1 | - | 2 - 3 | 625 | 85 | |
Set Membership: |
|