Search
Search

ccnd2a| Gene Symbol: ccnd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089445.1 | NP_001082914.1 | 2 | 549 | 96 | |
| XM_005174351.3 | XP_005174408.1 | 3 | 337 | 96 | ||
| GenBank mRNA | BC133931.1 | - | 2 | 549 | 96 | |
| FJ648073.1 | - | 2 | 560 | 96 | ||
Set Membership: |
|