Search
Search

efna2a| Gene Symbol: efna2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131022.2 | NP_571097.1 | 1 | 454 | 67 | |
| GenBank mRNA | BC080234.1 | - | 1 | 775 | 67 | |
| BC096783.1 | - | 1 | 454 | 67 | ||
| BC164994.1 | - | 1 | 97 | 67 | ||
| KM655628.1 | - | 1 | 63 | 67 | ||
Set Membership: |
|