Search
Search

cldna| Gene Symbol: cldna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131762.2 | NP_571837.2 | 1 | 1141 | 85 | |
| GenBank mRNA | AF359423.1 | - | 1 | 1141 | 85 | |
| BC065445.1 | - | 1 | 1141 | 85 | ||
Set Membership: |
|