Search
Search

pdxka| Gene Symbol: pdxka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126449.1 | NP_001119921.1 | 8 - 9 | 845 | 74 | |
| XM_021478775.1 | XP_021334450.1 | 8 - 9 | 837 | 74 | ||
| GenBank mRNA | BC085468.1 | - | 8 - 9 | 802 | 74 | |
| BC153430.1 | - | 8 - 9 | 799 | 74 | ||
| BC164257.1 | - | 8 - 9 | 645 | 74 | ||
Set Membership: |
|