Search
Search

ndufaf1| Gene Symbol: ndufaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013308.2 | NP_001013326.1 | 4 - 5 | 906 | 77 | |
| XM_005156828.4 | XP_005156885.1 | 2 - 3 | 754 | 77 | ||
| XM_005156829.4 | XP_005156886.1 | 4 - 5 | 932 | 77 | ||
| XM_005156830.4 | XP_005156887.1 | 3 - 4 | 870 | 77 | ||
| XM_005156831.4 | XP_005156888.1 | 3 - 4 | 873 | 77 | ||
| XM_005156832.4 | XP_005156889.1 | 2 - 3 | 807 | 77 | ||
| XM_009307315.3 | XP_009305590.1 | 4 - 5 | 980 | 77 | ||
| XM_017358738.2 | XP_017214227.1 | 3 - 4 | 927 | 77 | ||
| GenBank mRNA | BC090683.1 | - | 4 - 5 | 910 | 77 | |
Set Membership: |
|