Search
Search

aste1a| Gene Symbol: aste1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099430.2 | NP_001092900.1 | 2 - 3 | 1365 | 82 | |
| GenBank mRNA | BC163487.1 | - | 2 - 3 | 1359 | 82 | |
| BC163493.1 | - | 2 - 3 | 1359 | 82 | ||
Set Membership: |
|