Search
Search

ada2a| Gene Symbol: ada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033717.2 | NP_001028889.2 | 2 - 3 | 425 | 85 | |
| GenBank mRNA | AF384217.1 | - | 2 - 3 | 425 | 85 | |
| BC045839.1 | - | 2 - 3 | 400 | 85 | ||
Set Membership: |
|